Anti-CADM 4 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-5997R
BOSSBS-5997REA 725 EUR
BOSSBS-5997R
Anti-CADM 4 Rabbit Polyclonal Antibody
Antikörper
Involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CADM 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat