Anti-EXTL1 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-6005R
BOSSBS-6005REA 725 EUR
BOSSBS-6005R
Anti-EXTL1 Rabbit Polyclonal Antibody
Antikörper
EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and is responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: EXTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat