Anti-EXTL1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6005R-CY3
BOSSBS-6005R-CY3EA 900 EUR
BOSSBS-6005R-CY3
Anti-EXTL1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and is responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EXTL1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR