Anti-Folliculin Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6007R-A680
BOSSBS-6007R-A680EA 893 EUR
BOSSBS-6007R-A680
Anti-Folliculin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene is located within the Smith-Magenis syndrome region on chromosome 17. Mutations in this gene are associated with Birt-Hogg-Dube syndrome, which is characterised by fibrofolliculomas, renal tumors, lung cysts, and pneumothorax. Alternative splicing of this gene results in three transcript variants encoding different isoforms.

Type: Primary
Antigen: Folliculin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 65-165/579
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR