Anti-SMARCA1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6015R
BOSSBS-6015REA 725 EUR
BOSSBS-6015R
Anti-SMARCA1 Rabbit Polyclonal Antibody
Antikörper
SNF2L, also known as SMARCA1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1), SWI or ISWI, is a 1,054 amino acid protein that localizes to the nucleus and contains one helicase C-terminal domain, one helicase ATP-binding domain and two SANT domains. Expressed as multiple alternatively spliced isoforms, SNF2L exists as a component of the nucleosome-remodeling factor (NURF) complex where it helps to facilitate the ATP-dependent perturbation of chromatin structure and may also be involved in brain development and neurite outgrowth. The gene encoding SNF2L maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SMARCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR