Anti-SNF2L Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-6015R-CY5.5
BOSSBS-6015R-CY5.5EA 900 EUR
BOSSBS-6015R-CY5.5
Anti-SNF2L Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
SNF2L, also known as SMARCA1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1), SWI or ISWI, is a 1,054 amino acid protein that localizes to the nucleus and contains one helicase C-terminal domain, one helicase ATP-binding domain and two SANT domains. Expressed as multiple alternatively spliced isoforms, SNF2L exists as a component of the nucleosome-remodeling factor (NURF) complex where it helps to facilitate the ATP-dependent perturbation of chromatin structure and may also be involved in brain development and neurite outgrowth. The gene encoding SNF2L maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNF2L
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR