Anti-ACY1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6019R
BOSSBS-6019REA 725 EUR
BOSSBS-6019R
Anti-ACY1 Rabbit Polyclonal Antibody
Antikörper
Aminoacylase 1 is a cytosolic, homodimeric, zinc binding enzyme that catalyzes the hydrolysis of acylated L amino acids to L amino acids and acyl group, and has been postulated to function in the catabolism and salvage of acylated amino acids. ACY1 has been assigned to chromosome 3p21.1, a region reduced to homozygosity in small cell lung cancer (SCLC), and its expression has been reported to be reduced or undetectable in SCLC cell lines and tumors. The amino acid sequence of human aminoacylase 1 is highly homologous to the porcine counterpart, and ACY1 is the first member of a new family of zinc binding enzymes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ACY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR