Anti-BST1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6023R-A750
BOSSBS-6023R-A750EA 893 EUR
BOSSBS-6023R-A750
Anti-BST1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Synthesises the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger that elicits calcium release from intracellular stores. May be involved in pre-B-cell growth.

Type: Primary
Antigen: BST1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/318
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR