Anti-TCF3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-6066R-CY5
BOSSBS-6066R-CY5EA 1350 EUR
BOSSBS-6066R-CY5
Anti-TCF3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Heterodimers between TCF3 and tissue-specific basic helix-loop-helix (bHLH) proteins play major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Dimers bind DNA on E-box motifs: 5'-CANNTG-3'. Binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TCF3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR