Anti-NIT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6077R-A680
BOSSBS-6077R-A680EA 893 EUR
BOSSBS-6077R-A680
Anti-NIT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Play a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. it is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.

Type: Primary
Antigen: NIT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 111-210/327
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR