Anti-NIT1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-6077R-CY5.5
BOSSBS-6077R-CY5.5EA 900 EUR
BOSSBS-6077R-CY5.5
Anti-NIT1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Play a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. it is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NIT1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR