Anti-RHOBTB1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-6092R-CY5.5
BOSSBS-6092R-CY5.5EA 900 EUR
BOSSBS-6092R-CY5.5
Anti-RHOBTB1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
RHOBTB1 is a member of the Rho family of the small GTPase superfamily. Is made of a GTPase domain, a proline-rich region, a tandem of 2 BTB (broad complex, tramtrack, and bric-a-brac) domains, and a conserved C-terminal region. Plays a role in small GTPase-mediated signal transduction and the organization of the actin filament system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RHOBTB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR