Anti-TR1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-6117R-A647
BOSSBS-6117R-A647EA 915 EUR
BOSSBS-6117R-A647
Anti-TR1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
May inhibit NODAL and BMP signaling during neural patterning (By similarity). May be a tumor suppressor in brain cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR