Anti-TR1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-6117R-CY5
BOSSBS-6117R-CY5EA 922 EUR
BOSSBS-6117R-CY5
Anti-TR1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
May inhibit NODAL and BMP signaling during neural patterning (By similarity). May be a tumor suppressor in brain cancers.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat