Anti-ZNF431 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-6120R-A555
BOSSBS-6120R-A555EA 893 EUR
BOSSBS-6120R-A555
Anti-ZNF431 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ZNF431 belongs to the krueppel C2H2-type zinc-finger protein family and is probably involved in transcriptional regulation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZNF431
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human