Anti-ZNF431 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6120R-A680
BOSSBS-6120R-A680EA 893 EUR
BOSSBS-6120R-A680
Anti-ZNF431 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ZNF431 belongs to the krueppel C2H2-type zinc-finger protein family and is probably involved in transcriptional regulation.

Type: Primary
Antigen: ZNF431
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 371-470/576
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR