Anti-ARL3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-6162R-CY5.5
BOSSBS-6162R-CY5.5EA 900 EUR
BOSSBS-6162R-CY5.5
Anti-ARL3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Small GTP-binding protein which cycles between an inactive GDP-bound and an active GTP-bound form, and the rate of cycling is regulated by guanine nucleotide exchange factors (GEF) and GTPase-activating proteins (GAP). GTP-binding protein that does not act as an allosteric activator of the cholera toxin catalytic subunit. Requires assistance from GTPase-activating proteins (GAPs) like RP2 and PDE6D, in order to cycle between inactive GDP-bound and active GTP-bound forms. Required for normal cytokinesis and cilia signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR