Anti-Rab3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-6180R-A555
BOSSBS-6180R-A555EA 893 EUR
BOSSBS-6180R-A555
Anti-Rab3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has four known isoforms: 3A, 3B, 3C and 3D. These proteins are expressed in the highest concentration in the brain and endocrine tissues and are involved in regulated exocytosis of hormones and neurotransmitters. Specifically Rab3's are thought to play a role in presynaptic vesicle trafficking and priming.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Rab3A+Rab3B+Rab3C+Rab3D
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR