Anti-Renin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-6184R-A750
BOSSBS-6184R-A750EA 915 EUR
BOSSBS-6184R-A750
Anti-Renin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
Type: Primary
Antigen: Renin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-406/406
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Dog