Anti-TRAF3IP2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-6202R-A647
BOSSBS-6202R-A647EA 893 EUR
BOSSBS-6202R-A647
Anti-TRAF3IP2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Could be involved in the activation of both NF-kappa-B via a NF-kappa-B inhibitor kinase (IKK)-dependent mechanism and stress-activated protein kinase (SAPK)/JNK.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TRAF3IP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat