Anti-TRAF3IP2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6202R-CY3
BOSSBS-6202R-CY3EA 900 EUR
BOSSBS-6202R-CY3
Anti-TRAF3IP2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Could be involved in the activation of both NF-kappa-B via a NF-kappa-B inhibitor kinase (IKK)-dependent mechanism and stress-activated protein kinase (SAPK)/JNK.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRAF3IP2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR