Anti-NMT2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-6233R-HRP
BOSSBS-6233R-HRPEA 900 EUR
BOSSBS-6233R-HRP
Anti-NMT2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
NMT2 catalyzes the reaction of N-terminal myristoylation of many signaling proteins. It transfers myristic acid from myristoyl coenzyme A to the amino group of a protein's N-terminal glycine residue. several distinct NMTs exist, varying in apparent molecular weight and /or subcellular distribution.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: NMT2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR