Anti-LCMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6249R-A680
BOSSBS-6249R-A680EA 893 EUR
BOSSBS-6249R-A680
Anti-LCMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Methylates the carboxyl group of the C-terminal leucine residue of protein phosphatase 2A catalytic subunits to form alpha-leucine ester residues.

Type: Primary
Antigen: LCMT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 45-150/334
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR