Anti-MAPK8IP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6255R-A750
BOSSBS-6255R-A750EA 893 EUR
BOSSBS-6255R-A750
Anti-MAPK8IP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The JNK-interacting protein (JIP) group of scaffold proteins selectively mediates JNK signaling by aggregating specific components of the MAPK cascade to form a functional JNK signaling module. Required for JNK activation in response to excitotoxic stress. Cytoplasmic MAPK8IP1 causes inhibition of JNK-regulated activity by retaining JNK in the cytoplasm and inhibiting JNK phosphorylation of c-Jun. May also participate in ApoER2-specific reelin signaling. Directly, or indirectly, regulates GLUT2 gene expression and beta-cell function. Appears to have a role in cell signaling in mature and developing nerve terminals. May function as a regulator of vesicle transport, through interactions with the JNK-signaling components and motor proteins (By similarity). Functions as an anti-apoptotic protein and whose level seems to influence the beta-cell death or survival response.

Type: Primary
Antigen: MAPK8IP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 425-524/711
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR