Anti-OSM Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-6295R-CY5
BOSSBS-6295R-CY5EA 900 EUR
BOSSBS-6295R-CY5
Anti-OSM Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Growth regulator. Inhibits the proliferation of a number of tumor cell lines. Stimulates proliferation of AIDS-KS cells. It regulates cytokine production, including IL-6, G-CSF and GM-CSF from endothelial cells. Uses both type I OSM receptor (heterodimers composed of LIPR and IL6ST) and type II OSM receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OSM
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR