Anti-4-HNE Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-6313R-A555
BOSSBS-6313R-A555EA 890 EUR
BOSSBS-6313R-A555
Anti-4-HNE Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Aldehydic products of lipid peroxidation, such as 4 hydroxynonenal (4 HNE), have been implicated in the etiology of pathological changes under oxidative stress as a key mediator of oxidative stress induced cell death. It is a stable product of lipid peroxidation, is proarrhythmic and may contribute to the cytotoxic effects of oxidative stress

4-HNE has been hypothesized to play a key role in cell signal transduction, in a variety of pathways from cell cycle events to cellular adhesion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 4-HNE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR