Anti-4 Hydroxynonenal Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-6313R-A680
BOSSBS-6313R-A680EA 890 EUR
BOSSBS-6313R-A680
Anti-4 Hydroxynonenal Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Aldehydic products of lipid peroxidation, such as 4 hydroxynonenal (4 HNE), have been implicated in the etiology of pathological changes under oxidative stress as a key mediator of oxidative stress induced cell death. It is a stable product of lipid peroxidation, is proarrhythmic and may contribute to the cytotoxic effects of oxidative stress
4-HNE has been hypothesised to play a key role in cell signal transduction, in a variety of pathways from cell cycle events to cellular adhesion.
Type: Primary
Antigen: 4 Hydroxynonenal
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Others; Monkey