Anti-4 Hydroxynonenal Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6313R-A750
BOSSBS-6313R-A750EA 890 EUR
BOSSBS-6313R-A750
Anti-4 Hydroxynonenal Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Aldehydic products of lipid peroxidation, such as 4 hydroxynonenal (4 HNE), have been implicated in the etiology of pathological changes under oxidative stress as a key mediator of oxidative stress induced cell death. It is a stable product of lipid peroxidation, is proarrhythmic and may contribute to the cytotoxic effects of oxidative stress

4-HNE has been hypothesised to play a key role in cell signal transduction, in a variety of pathways from cell cycle events to cellular adhesion.

Type: Primary
Antigen: 4 Hydroxynonenal
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Others; Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR