Anti-4-HNE Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-6313R-CY3
BOSSBS-6313R-CY3EA 897 EUR
BOSSBS-6313R-CY3
Anti-4-HNE Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Aldehydic products of lipid peroxidation, such as 4 hydroxynonenal (4 HNE), have been implicated in the etiology of pathological changes under oxidative stress as a key mediator of oxidative stress induced cell death. It is a stable product of lipid peroxidation, is proarrhythmic and may contribute to the cytotoxic effects of oxidative stress
4-HNE has been hypothesized to play a key role in cell signal transduction, in a variety of pathways from cell cycle events to cellular adhesion.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: 4-HNE
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others