Anti-ALOX15B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-6335R-A750
BOSSBS-6335R-A750EA 893 EUR
BOSSBS-6335R-A750
Anti-ALOX15B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Non-heme iron-containing dioxygenase that catalyzes the stereo-specific peroxidation of free and esterified polyunsaturated fatty acids generating a spectrum of bioactive lipid mediators. Converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid/(15S)-HPETE. Also acts on linoleic acid to produce 13-hydroxyoctadecadienoic acid/13-HPODE. Has no detectable 8S-lipoxygenase activity but reacts with (8S)-HPETE to produce (8S,15S)-diHPETE. May regulate progression through the cell cycle and cell proliferation. May also regulate cytokine secretion by macrophages and therefore play a role in the immune response. May also regulate macrophage differentiation into proatherogenic foam cells.
Type: Primary
Antigen: ALOX15B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 13-110/676
Host: Rabbit
Isotype: IgG
Reactivity: Rat