Anti-Adenylosuccinate Lyase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-6352R-A750
BOSSBS-6352R-A750EA 893 EUR
BOSSBS-6352R-A750
Anti-Adenylosuccinate Lyase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Adenylsuccinate lyase is involved in both de novo synthesis of purines and formation of adenosine monophosphate from inosine monophosphate. It catalyzes two reactions in AMP biosynthesis: the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and removal of fumarate from adenylosuccinate to give AMP. Adenylosuccinase deficiency results in succinylpurinemic autism, psychomotor retardation, and , in some cases, growth retardation associated with muscle wasting and epilepsy. Two transcript variants encoding different isoforms have been found for this gene.
Type: Primary
Antigen: Adenylosuccinate Lyase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 185-280/484
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse