Anti-p70 S6 kinase alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-6370R-A750
BOSSBS-6370R-A750EA 890 EUR
BOSSBS-6370R-A750
Anti-p70 S6 kinase alpha Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Serine/threonine-protein kinase that acts downstream of mTOR signaling in response to growth factors and nutrients to promote cell proliferation, cell growth and cell cycle progression. Regulates protein synthesis through phosphorylation of EIF4B, RPS6 and EEF2K, and contributes to cell survival by repressing the pro-apoptotic function of BAD. Under conditions of nutrient depletion, the inactive form associates with the EIF3 translation initiation complex. Upon mitogenic stimulation, phosphorylation by the mammalian target of rapamycin complex 1 (mTORC1) leads to dissociation from the EIF3More Product Information