Anti-UHRF2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6389R
BOSSBS-6389REA 725 EUR
BOSSBS-6389R
Anti-UHRF2 Rabbit Polyclonal Antibody
Antikörper
E3 ubiquitin-protein ligase which mediates ubiquitination and subsequent proteasomal degradation of PCNP. May participate in methylation-dependent transcriptional regulation. Important for G1/S transition. Overexpression causes G1 phase cell arrest.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UHRF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR