Anti-TN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-6413R-A488
BOSSBS-6413R-A488EA 893 EUR
BOSSBS-6413R-A488
Anti-TN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Tetranectin binds to plasminogen and to isolated kringle 4. May be involved in the packaging of molecules destined for exocytosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Tetranectin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat