Anti-CATSPER4 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-6422R-CY5.5
BOSSBS-6422R-CY5.5EA 900 EUR
BOSSBS-6422R-CY5.5
Anti-CATSPER4 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
CatSpers (cation channel, sperm associated proteins) are ion transport proteins located on the surface of sperm cells in the principal piece of the sperm tail. CatSpers are vital to sperm motility, fertilization and cAMP-mediated calcium influx in sperm. There are four CatSper proteins in mammalian sperm, namely CatSper (or CatSper1), CatSper2, CatSper3 and CatSper4. CatSper proteins contain a single, six-transmembrane-spanning segment and exhibit the voltage-dependent Ca2+ channel four-repeat structure. CatSper proteins are believed to assemble into a heterotetrameric complex, forming anMore Product Information