Anti-UHRF1/2 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-6427R
BOSSBS-6427REA 757 EUR
BOSSBS-6427R
Anti-UHRF1/2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UHRF1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat