Anti-HNRNPC Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-6459R-CY7
BOSSBS-6459R-CY7EA 900 EUR
BOSSBS-6459R-CY7
Anti-HNRNPC Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Binds pre-mRNA and nucleates the assembly of 40S hnRNP particles. Single HNRNPC tetramers bind 230-240 nucleotides. Trimers of HNRNPC tetramers bind 700 nucleotides. May play a role in the early steps of spliceosome assembly and pre-mRNA splicing. Interacts with poly-U tracts in the 3'-UTR or 5'-UTR of mRNA and modulates the stability and the level of translation of bound mRNA molecules.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HNRNPC
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat