Anti-DLX1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-6467R-CY5
BOSSBS-6467R-CY5EA 900 EUR
BOSSBS-6467R-CY5
Anti-DLX1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Likely to play a regulatory role in the development of the ventral forebrain. May play a role in craniofacial patterning and morphogenesis and may be involved in the early development of diencephalic subdivisions.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DLX1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat