Anti-CKAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-6520R-A350
BOSSBS-6520R-A350EA 893 EUR
BOSSBS-6520R-A350
Anti-CKAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
High-affinity epithelial cell surface receptor for APF. Mediates the anchoring of the endoplasmic reticulum to microtubules.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CKAP4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR