Anti-CKAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6520R-A680
BOSSBS-6520R-A680EA 893 EUR
BOSSBS-6520R-A680
Anti-CKAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
High-affinity epithelial cell surface receptor for APF. Mediates the anchoring of the endoplasmic reticulum to microtubules.

Type: Primary
Antigen: CKAP4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 525-602/602
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR