Anti-CKAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-6520R-A750
BOSSBS-6520R-A750EA 893 EUR
BOSSBS-6520R-A750
Anti-CKAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
High-affinity epithelial cell surface receptor for APF. Mediates the anchoring of the endoplasmic reticulum to microtubules.
Type: Primary
Antigen: CKAP4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 525-602/602
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat