Anti-CKAP4 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6520R-CY3
BOSSBS-6520R-CY3EA 900 EUR
BOSSBS-6520R-CY3
Anti-CKAP4 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
High-affinity epithelial cell surface receptor for APF. Mediates the anchoring of the endoplasmic reticulum to microtubules.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CKAP4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

TECHNISCHE DATEN

Antigen-Symbol CKAP4
Environmentally Preferable
Antigen-Name Cytoskeleton-associated protein 4
Antigen-Synonyme CLIMP-63|p63|ERGIC-63
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy3®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 10970
Isotyp IgG
ImmunoChemie Yes
Immunogen 552-602/602
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR