Anti-CKAP4 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-6520R-CY5.5
BOSSBS-6520R-CY5.5EA 900 EUR
BOSSBS-6520R-CY5.5
Anti-CKAP4 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
High-affinity epithelial cell surface receptor for APF. Mediates the anchoring of the endoplasmic reticulum to microtubules.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CKAP4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen