Anti-CKAP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-6520R-HRP
BOSSBS-6520R-HRPEA 900 EUR
BOSSBS-6520R-HRP
Anti-CKAP4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
High-affinity epithelial cell surface receptor for APF. Mediates the anchoring of the endoplasmic reticulum to microtubules.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CKAP4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR