Anti-CD39L4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-6526R-A750
BOSSBS-6526R-A750EA 893 EUR
BOSSBS-6526R-A750
Anti-CD39L4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Uridine diphosphatase (UDPase) that promotes protein N-glycosylation and ATP level regulation. UDP hydrolysis promotes protein N-glycosylation and folding in the endoplasmic reticulum, as well as elevated ATP consumption in the cytosol via an ATP hydrolysis cycle. Together with CMPK1 and AK1, constitutes an ATP hydrolysis cycle that converts ATP to AMP and results in a compensatory increase in aerobic glycolysis. The nucleotide hydrolyzing preference is GDP > IDP > UDP, but not any other nucleoside di-, mono- or triphosphates, nor thiamine pyrophosphate. Plays a key role in the AKT1-PTEN signaling pathway by promoting glycolysis in proliferating cells in response to phosphoinositide 3-kinase (PI3K) signaling.
Type: Primary
Antigen: CD39L4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-380/428
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen