Anti-GPAT3 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6547R
BOSSBS-6547REA 725 EUR
BOSSBS-6547R
Anti-GPAT3 Rabbit Polyclonal Antibody
Antikörper
May transfer the acyl-group from acyl-coA to the sn-1 position of glycerol-3-phosphate, an essential step in glycerolipid biosynthesis. Also transfers the acyl-group from acyl-coA to the sn-2 position of 1-acyl-sn-glycerol-3-phosphate (lysophosphatidic acid, or LPA), forming 1,2-diacyl-sn-glycerol-3-phosphate (phosphatidic acid, or PA).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR