Anti-MMTN Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-6551R-A647
BOSSBS-6551R-A647EA 893 EUR
BOSSBS-6551R-A647
Anti-MMTN Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene cause progressive encephalopathy resulting from mitochondrial complex I deficiency.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMTN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR