Anti-Mimitin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6551R-A750
BOSSBS-6551R-A750EA 893 EUR
BOSSBS-6551R-A750
Anti-Mimitin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene cause progressive encephalopathy resulting from mitochondrial complex I deficiency.

Type: Primary
Antigen: Mimitin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 71-169/169
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR