Anti-RNASE4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6583R-A680
BOSSBS-6583R-A680EA 893 EUR
BOSSBS-6583R-A680
Anti-RNASE4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This RNase has marked specificity towards the 3' side of uridine nucleotides.

Type: Primary
Antigen: RNASE4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-147/147
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR