Anti-HSD17B6 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6592R
BOSSBS-6592REA 725 EUR
BOSSBS-6592R
Anti-HSD17B6 Rabbit Polyclonal Antibody
Antikörper
NAD-dependent oxidoreductase with broad substrate specificity that shows both oxidative and reductive activity (in vitro). Has 17-beta-hydroxysteroid dehydrogenase activity towards various steroids (in vitro). Converts 5-alpha-androstan-3-alpha,17-beta-diol to androsterone and estradiol to estrone (in vitro). Has 3-alpha-hydroxysteroid dehydrogenase activity towards androsterone (in vitro). Has retinol dehydrogenase activity towards all-trans-retinol (in vitro). Can convert androsterone to epi-androsterone. Androsterone is first oxidized to 5-alpha-androstane-3,17-dione and then reduced toMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol HSD17B6
Environmentally Preferable
Antigen-Name Hydroxysteroid (17-beta) Dehydrogenase 6
Antigen-Synonyme SDR9C6|HSE|RODH
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 8630
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Form liquid
Reinigung Purified by Protein A.
Konzentration 1 ug/ul
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transporttemperatur 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR